Learn More
Cayman Chemical N,N'-Diphenethylurea, Purity: ≥95%, CAS: 5467-84-5, 10 mg
SDP
Catalog No. 502947094
Encompass_Preferred
A bacterial metabolite with diverse biological activities; inhibits Streptomyces chitin synthase 1 (IC50 = 14 µg/ml); active against C. neoformans and R. solani (MIC = 50 µg/ml for both); enhances insulin-induced adipocyte differentiation in 3T3-L1 and C3H/10T1/2 cells at 30 and 100 µM
Catalog No. 50-294-7094 Supplier Cayman Chemical Supplier No. 3358010 MG
A bacterial metabolite with diverse biological activities; inhibits Streptomyces chitin synthase 1 (IC50 = 14 µg/ml); active against C. neoformans and R. solani (MIC = 50 µg/ml for both); enhances insulin-induced adipocyte differentiation in 3T3-L1 and C3H/10T1/2 cells at 30 and 100 µM