Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Diphenyl-p-phenylenediamine, 97%

Catalog No. p-7027167
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2371306 5 g
AAB2371314 25 g
AAB2371322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2371306 Supplier Thermo Scientific Chemicals Supplier No. B2371306
Only null left
Add to Cart

CAS: 74-31-7 | C18H16N2 | 260.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-31-7
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00003015
InChI Key UTGQNNCQYDRXCH-UHFFFAOYSA-N
Synonym n,n'-diphenyl-p-phenylenediamine, n1,n4-diphenylbenzene-1,4-diamine, n,n'-diphenyl-1,4-phenylenediamine, dppd, flexamine g, agerite dppd, diphenyl-p-phenylenediamine, dffd, nonflex h, stabilizer dppd
PubChem CID 6319
ChEBI CHEBI:34860
IUPAC Name 1-N,4-N-diphenylbenzene-1,4-diamine
SMILES N(C1=CC=CC=C1)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 146°C to 153°C
Density 1.28
Boiling Point 220°C to 225°C (1 mmHg)
Quantity 5 g
Merck Index 14,3329
Formula Weight 260.34
Percent Purity 97%
Chemical Name or Material N,N'-Diphenyl-p-phenylenediamine
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 200-806-4
RTECSNumber ST2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.