Learn More
Medchemexpress LLC N,N'-Diphenylthiourea | 102-08-9 | 99.05% | 228.32 | 250 G
SDP
Catalog No. 5000453310
Encompass_Preferred
N,N′-Diphenylthiourea is a biological material or organic compound suitable for life science research. It is provided as a solid, white to off-white in appearance, with a high purity of 99.05%. This product is for research use only and not sold to patients.
  • Used in life science research
  • Solid appearance
  • White to off-white color
  • Stable under recommended storage conditions
  • Recommended storage at -20°C for 3 years or 4°C for 2 years (powder)
  • Recommended storage at -80°C for 6 months or -20°C for 1 month (in solvent)
Catalog No. 50-004-53310 Supplier Medchemexpress LLC Supplier No. 250GHYW012276
N,N′-Diphenylthiourea is a biological material or organic compound suitable for life science research. It is provided as a solid, white to off-white in appearance, with a high purity of 99.05%. This product is for research use only and not sold to patients.
  • Used in life science research
  • Solid appearance
  • White to off-white color
  • Stable under recommended storage conditions
  • Recommended storage at -20°C for 3 years or 4°C for 2 years (powder)
  • Recommended storage at -80°C for 6 months or -20°C for 1 month (in solvent)