Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dipropyl-L-alanine 98.0+%, TCI America™
SDP

Catalog No. D22241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22241G Supplier TCI America Supplier No. D22241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81854-56-0
Molecular Formula C9H19NO2
Molecular Weight (g/mol) 173.256
MDL Number MFCD00010823
InChI Key GZRZQYMPUHUFSQ-QMMMGPOBSA-N
PubChem CID 14274716
IUPAC Name (2S)-2-(dipropylamino)propanoic acid
SMILES CCCN(CCC)C(C)C(=O)O

Specifications

Melting Point 96°C
Color White
Quantity 1 g
Formula Weight 173.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dipropyl-L-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.