Learn More
Medchemexpress LLC N,N'-DME-N-PEG2-Boc | 2279944-69-1 | >98.0% | 304.43 | 250 MG
SDP
Catalog No. 5000351571
Encompass_Preferred
N,N'-DME-N-PEG2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is recommended to store this product at 4°C, protected from light. If dissolved in a solvent, store at -80°C for up to 6 months or -20°C for 1 month, also protected from light.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
Catalog No. 50-003-51571 Supplier Medchemexpress LLC Supplier No. 250MGHY140163
N,N'-DME-N-PEG2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is recommended to store this product at 4°C, protected from light. If dissolved in a solvent, store at -80°C for up to 6 months or -20°C for 1 month, also protected from light.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs