Learn More
Cayman Chemical N,N-DMT N-oxide, Purity: ≥98%, CAS: 948-19-6, 1 mg
SDP
Catalog No. 502949507
Encompass_Preferred
An analytical reference standard categorized as a tryptamine metabolite; a metabolite of N,N-DMT; intended for research and forensic applications
Catalog No. 50-294-9507 Supplier Cayman Chemical Supplier No. 335541 MG
An analytical reference standard categorized as a tryptamine metabolite; a metabolite of N,N-DMT; intended for research and forensic applications