Learn More
Cayman Chemical N,N-DMT N-oxide, Purity: ≥98%, CAS: 948-19-6, 5 mg
SDP
Catalog No. 502949508
Encompass_Preferred
An analytical reference standard categorized as a tryptamine metabolite; a metabolite of N,N-DMT; intended for research and forensic applications
Catalog No. 50-294-9508 Supplier Cayman Chemical Supplier No. 335545 MG
An analytical reference standard categorized as a tryptamine metabolite; a metabolite of N,N-DMT; intended for research and forensic applications