Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-n-Hexyl-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH58403MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58403MF 50 mg
AAH58403MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH58403MF Supplier Thermo Scientific Chemicals Supplier No. H58403MF

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52374-18-2
Molecular Formula C12H18N2O4S
Molecular Weight (g/mol) 286.346
MDL Number MFCD00438183
InChI Key HMKTZTGMSGRPHW-UHFFFAOYSA-N
Synonym n-hexyl-4-nitro-benzenesulfonamide, hexyl 4-nitrophenyl sulfonyl amine, n-n-hexyl-4-nitrobenzenesulfonamide, 4-nitro-benzenesulfonic acid hexylamide
PubChem CID 296253
IUPAC Name N-hexyl-4-nitrobenzenesulfonamide
SMILES CCCCCCNS(=O)(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 50 mg
Beilstein 2667220
Formula Weight 286.34
Percent Purity 97%
Chemical Name or Material N-n-Hexyl-4-nitrobenzenesulfonamide
Hazard Category H225-H261-H314-H351-H335
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.