Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Methylenebisacrylamide, ≥99.9%, Molecular Biology Grade, Ultrapure, Thermo Scientific™

Catalog No. p-7082853
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle

Catalog No. AAJ75821A1

Supplier: Thermo Scientific J75821A1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Cross-linking agent used in forming polyacrylamide gels
  • Porosity of gel is determined by chain length and degree of cross-linking
  • Most sequencing gels use ratio of 1 molecule of cross-linker for every 19 monomers of acrylamide
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym n,n'-methylenebisacrylamide, n,n'-methylenediacrylamide, n,n'-methylene-bis-acrylamide, methylenebisacrylamide, methylenediacrylamide, 2-propenamide, n,n'-methylenebis, bis-acrylamide, n,n'-methylenebis acrylamide, n,n'-methylidenebisacrylamide, bisacrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Melting Point >300°C
Density 1.235
Packaging Plastic bottle
Linear Formula (CH2=CHCONH)2CH2
Quantity 1 kg
Beilstein 1706297
Formula Weight 154.17
Percent Purity ≥99.9%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material N,N'-Methylenebisacrylamide
Show More Show Less
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 203-750-9
RTECSNumber AS3678000
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.