Learn More
N,N'-Methylenebisacrylamide, ≥99.9%, Molecular Biology Grade, Ultrapure, Thermo Scientific™
Shop All Thermo Scientific ProductsDescription
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Cross-linking agent used in forming polyacrylamide gels
- Porosity of gel is determined by chain length and degree of cross-linking
- Most sequencing gels use ratio of 1 molecule of cross-linker for every 19 monomers of acrylamide
Specifications
Specifications
| Melting Point | >300°C |
| Density | 1.235 |
| Packaging | Plastic bottle |
| Linear Formula | (CH2=CHCONH)2CH2 |
| Quantity | 1 kg |
| Beilstein | 1706297 |
| Formula Weight | 154.17 |
| Percent Purity | ≥99.9% |
| Grade | Molecular Biology |
| Physical Form | Crystalline Powder |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 203-750-9 |
| RTECSNumber | AS3678000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.