Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N″,N″-Pentakis(2-hydroxypropyl)diethylenetriamine 95.0+%, TCI America™
SDP

Catalog No. P083225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P083225G 25 g
P0832100G 100 g
P0832500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P083225G Supplier TCI America Supplier No. P083225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17121-34-5
Molecular Formula C19H43N3O5
Molecular Weight (g/mol) 393.569
MDL Number MFCD00012064
InChI Key LWMWZNOCSCPBCH-UHFFFAOYSA-N
PubChem CID 86512
IUPAC Name 1-[bis[2-[bis(2-hydroxypropyl)amino]ethyl]amino]propan-2-ol
SMILES CC(CN(CCN(CC(C)O)CC(C)O)CCN(CC(C)O)CC(C)O)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 393.57
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′′,N′′-Pentakis(2-hydroxypropyl)diethylenetriamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.