Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetra(2-naphthyl)benzidine 98.0+%, TCI America™
SDP

Catalog No. T32881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32881G Supplier TCI America Supplier No. T32881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141752-82-1
Molecular Formula C52H36N2
Molecular Weight (g/mol) 688.87
MDL Number MFCD03093244
InChI Key QKCGXXHCELUCKW-UHFFFAOYSA-N
PubChem CID 21881240
IUPAC Name N4,N4,N4',N4'-tetrakis(naphthalen-2-yl)-[1,1'-biphenyl]-4,4'-diamine
SMILES C1=CC2=CC=C(C=C2C=C1)N(C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 171°C
Color Yellow
Quantity 1 g
Formula Weight 688.87
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetra(2-naphthyl)benzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.