Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N,N',N'-Tetraacetylethylenediamine

Catalog No. 8654550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-545-50G 50 g
86-545-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 86-545-50G Supplier Sigma Aldrich Supplier No. 8654550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N, n, nα, nα-tetraacetylethylenediamine (taed) can be used as a reagent:
  • Along with sodium perborate and sodium bicarbonate for the oxidation of primary aliphatic amines to aliphatic c-nitroso compounds
  • In combination with sodium percarbonate to deliver peracetic acid in situ for baeyer-villiger oxidation.it can also be used as a peroxide bleach activator in laundry detergents and in textile chemistry applications

Specifications

CAS 10543-57-4
Molecular Formula C10H16N2O4
Linear Formula C10H16N2O4
MDL Number MFCD00014967
Quantity 50 g
Synonym TAED; N,N-Ethylenebis(diacetamide)
Molecular Weight (g/mol) 228.25
Percent Purity ≥90% (CHN)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.