Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetrabutylmalonamide 97.0+%, TCI America™
SDP

Catalog No. T19685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19685G Supplier TCI America Supplier No. T19685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14287-98-0
Molecular Formula C19H38N2O2
Molecular Weight (g/mol) 326.53
MDL Number MFCD00089288
InChI Key QTVRIQFMPJRJAK-UHFFFAOYSA-N
PubChem CID 3494509
IUPAC Name N,N,N',N'-tetrabutylpropanediamide
SMILES CCCCN(CCCC)C(=O)CC(=O)N(CCCC)CCCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 326.53
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetrabutylmalonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.