Learn More
Medchemexpress LLC N,N,N',N'-tetracyclohexyl-3-oxapentanediamide | 74267-27-9 | ≥98.0% | 460.69 g·mol⁻¹ | C28H48N2O3 | 5 MG
SDP
Catalog No. 5000223272
Encompass_Preferred
N,N,N',N'-tetracyclohexyl-3-oxapentanediamide is a lipophilic ionophore used to facilitate selective calcium transport in analytical and sensor applications. It is provided as a powder for research use and is commonly employed in the preparation of calcium ion-selective electrodes and related assays.
  • Lipophilic calcium ionophore for selective Ca2+ transport.
  • Intended for preparation of calcium ion-selective electrodes.
  • Supplied as a white to off-white powder suitable for analytical use.
  • High purity supports reproducible analytical performance.
  • Molecular weight 460.69 g·mol⁻¹ and formula C28H48N2O3 aid in formulation and calculation.
  • Stable under recommended low-temperature storage conditions.
Catalog No. 50-002-23272 Supplier Medchemexpress LLC Supplier No. HY1397285MG
N,N,N',N'-tetracyclohexyl-3-oxapentanediamide is a lipophilic ionophore used to facilitate selective calcium transport in analytical and sensor applications. It is provided as a powder for research use and is commonly employed in the preparation of calcium ion-selective electrodes and related assays.
  • Lipophilic calcium ionophore for selective Ca2+ transport.
  • Intended for preparation of calcium ion-selective electrodes.
  • Supplied as a white to off-white powder suitable for analytical use.
  • High purity supports reproducible analytical performance.
  • Molecular weight 460.69 g·mol⁻¹ and formula C28H48N2O3 aid in formulation and calculation.
  • Stable under recommended low-temperature storage conditions.