Learn More
Medchemexpress LLC N,n,n′,n′-tetracyclohexyl-3-oxapentanediamide | 74267-27-9 | 99.4% | 460.69 | 250 MG
SDP
Catalog No. 5000351570
Encompass_Preferred
N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) is a lipophilic ionophore used in preparing calcium ion-selective electrodes for research applications. This compound can be utilized to measure intracellular free calcium.
  • Used in calcium ion-selective electrodes
  • Measures intracellular free calcium
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-51570 Supplier Medchemexpress LLC Supplier No. 250MGHY139728
N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) is a lipophilic ionophore used in preparing calcium ion-selective electrodes for research applications. This compound can be utilized to measure intracellular free calcium.
  • Used in calcium ion-selective electrodes
  • Measures intracellular free calcium
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month