Learn More
Medchemexpress LLC N,N,N',N'-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) | 74267-27-9 | 99.4% | 460.69 | 100 MG
SDP
Catalog No. 5000351518
Encompass_Preferred
N,N,N',N'-Tetracyclohexyl-3-oxapentanediamide, also known as Calcium ionophore II, is a lipophilic ionophore. It is commonly used in preparing calcium ion-selective electrodes due to its ability to facilitate the transport of calcium ions across membranes. This chemical has a high purity of 99.36% and a molecular weight of 460.69.
  • High purity for accurate research
  • Functions as a lipophilic ionophore
  • Essential for creating calcium ion-selective electrodes
  • Facilitates efficient calcium ion transport
Catalog No. 50-003-51518 Supplier Medchemexpress LLC Supplier No. HY139728100MG
N,N,N',N'-Tetracyclohexyl-3-oxapentanediamide, also known as Calcium ionophore II, is a lipophilic ionophore. It is commonly used in preparing calcium ion-selective electrodes due to its ability to facilitate the transport of calcium ions across membranes. This chemical has a high purity of 99.36% and a molecular weight of 460.69.
  • High purity for accurate research
  • Functions as a lipophilic ionophore
  • Essential for creating calcium ion-selective electrodes
  • Facilitates efficient calcium ion transport