Learn More
Medchemexpress LLC N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) | 74267-27-9 | 99.4% | 460.69 | 25 MG
SDP
Catalog No. 5000348716
Encompass_Preferred
N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) is a lipophilic ionophore that can be used in preparing calcium ion-selective electrodes. Calcium ion-selective electrodes can be used to measure intracellular free calcium.
  • Lipophilic ionophore
  • Used in preparing calcium ion-selective electrodes
  • Enables measurement of intracellular free calcium
  • For research use only
Catalog No. 50-003-48716 Supplier Medchemexpress LLC Supplier No. 25MGHY139728
N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) is a lipophilic ionophore that can be used in preparing calcium ion-selective electrodes. Calcium ion-selective electrodes can be used to measure intracellular free calcium.
  • Lipophilic ionophore
  • Used in preparing calcium ion-selective electrodes
  • Enables measurement of intracellular free calcium
  • For research use only