Learn More
Medchemexpress LLC N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) | 74267-27-9 | 99.4% | 460.69 | 500 MG
SDP
Catalog No. 5000349878
Encompass_Preferred
N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) is a lipophilic ionophore that can be used in preparing calcium ion-selective electrodes. Calcium ion-selective electrodes can be used to measure intracellular free calcium.
  • Lipophilic ionophore
  • Used in preparing calcium ion-selective electrodes
  • Calcium ion-selective electrodes measure intracellular free calcium
Catalog No. 50-003-49878 Supplier Medchemexpress LLC Supplier No. HY139728500MG
N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide (Calcium ionophore II) is a lipophilic ionophore that can be used in preparing calcium ion-selective electrodes. Calcium ion-selective electrodes can be used to measure intracellular free calcium.
  • Lipophilic ionophore
  • Used in preparing calcium ion-selective electrodes
  • Calcium ion-selective electrodes measure intracellular free calcium