Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%

Catalog No. AC420485000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC420485000 500 g Glass bottle
AC420481000 100 g Glass bottle
AC420480010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC420485000 Supplier Thermo Scientific Chemicals Supplier No. 420485000
  • $168.00
    $144.65 / Each of 1

    Save $23.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 140-07-8 | C10H24N2O4 | 236.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-07-8
Molecular Formula C10H24N2O4
Molecular Weight (g/mol) 236.31
MDL Number MFCD00002854
InChI Key BYACHAOCSIPLCM-UHFFFAOYSA-N
Synonym theed, n,n,n',n'-tetrakis 2-hydroxyethyl ethylenediamine, ethylenediamine tetraethanol, entol, tked, ethylenedinitrilotetraethanol, tetrakis hydroxyethyl ethylenediamine, tetra hydroxyethyl ethylenediamine, unii-k5ape098zi, tetrahydroxyethyl ethylenediamine
PubChem CID 67322
SMILES OCCN(CCO)CCN(CCO)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Amber
Density 1.1300g/mL
pH ≥10 (25% soln.)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4990 to 1.5070
Linear Formula (HOCH2CH2)2NCH2CH2N(CH2CH2OH)2
Quantity 500 g
Specific Gravity 1.13
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in ethanol
IUPAC Name 2-({2-[bis(2-hydroxyethyl)amino]ethyl}(2-hydroxyethyl)amino)ethan-1-ol
Formula Weight 236.31
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
Show More Show Less
EINECSNumber 205-396-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.