Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine 97.0+%, TCI America™
SDP

Catalog No. T14875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14875G 5 g
T14871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14875G Supplier TCI America Supplier No. T14875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16858-02-9 | C26H28N6 | 424.55 g/mol

Chemical Identifiers

CAS 16858-02-9
Molecular Formula C26H28N6
Molecular Weight (g/mol) 424.55
MDL Number MFCD00036918
InChI Key CVRXLMUYFMERMJ-UHFFFAOYSA-N
Synonym tpen, n,n,n',n'-tetrakis 2-pyridylmethyl ethylenediamine, n1,n1,n2,n2-tetrakis pyridin-2-ylmethyl ethane-1,2-diamine, unii-r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridylmethyl ethane-1,2-diamine, r9ptu1u29i, n,n,n',n'-tetrakis 2-pyridyl-methyl ethylenediamine, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl-, oc-6-11, 1,2-ethanediamine,n,n,n',n'-tetrakis 2-pyridinylmethyl, 1,2-ethanediamine, n,n,n',n'-tetrakis 2-pyridinylmethyl
PubChem CID 5519
IUPAC Name (2-{bis[(pyridin-2-yl)methyl]amino}ethyl)bis[(pyridin-2-yl)methyl]amine
SMILES C(CN(CC1=CC=CC=N1)CC1=CC=CC=N1)N(CC1=CC=CC=N1)CC1=CC=CC=N1

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 424.55
Percent Purity ≥97.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.