Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetrakis(p-tolyl)benzidine 98.0+%, TCI America™
SDP

Catalog No. T22695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22695G Supplier TCI America Supplier No. T22695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76185-65-4
Molecular Formula C40H36N2
Molecular Weight (g/mol) 544.742
MDL Number MFCD00799300
InChI Key MVIXNQZIMMIGEL-UHFFFAOYSA-N
Synonym N,N,N′C,N′C-Tetrakis(4-methylphenyl)benzidine
PubChem CID 9850394
IUPAC Name 4-methyl-N-[4-[4-(4-methyl-N-(4-methylphenyl)anilino)phenyl]phenyl]-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C4=CC=C(C=C4)N(C5=CC=C(C=C5)C)C6=CC=C(C=C6)C

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 5 g
Formula Weight 544.74
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′,N′-Tetrakis(p-tolyl)benzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.