Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-1,3-diaminobutane 98.0+%, TCI America™
SDP

Catalog No. T0524500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0524500ML 500 mL
T052425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0524500ML Supplier TCI America Supplier No. T0524500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97-84-7
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00025678
InChI Key AXFVIWBTKYFOCY-UHFFFAOYNA-N
Synonym 1,3-bis dimethylamino butane, n,n,n',n'-tetramethyl-1,3-butanediamine, 1,3-butanediamine, n,n,n',n'-tetramethyl, tetramethylbutanediamine, tetramethylbutane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminobutane, niax catalyst tmbda, tetramethyl butanediamine, n,n,n',n'-tetramethylbutane-1,3-diamine, n,n,n1,n1-tetramethyl-1,3-diaminobutane
PubChem CID 7350
IUPAC Name [3-(dimethylamino)butyl]dimethylamine
SMILES CC(CCN(C)C)N(C)C

Specifications

Color Yellow
Boiling Point 169°C
Quantity 500 mL
UN Number 2733
Formula Weight 144.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,3-diaminobutane
RTECSNumber EJ7525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.