Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N,N',N'-Tetramethyl-1,3-propanediamine

Catalog No. 549983500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-998-3500ML 500 mL
54-998-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-998-3500ML Supplier Sigma Aldrich Supplier No. 549983500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n,nα,nα-tetramethyl-1,3-propanediamine is an acyclic tertiary amine
  • It shows the ability to absorb co2 when dissolved in aqueous solution.product of arkema inc

Specifications

CAS 110-95-2
Density 0.779 g/mL (at 25°C (literature))
Boiling Point 145°C to 146°C (lit.)
Molecular Formula C7H18N2
Refractive Index n20/D 1.4234 (literature)
Linear Formula (CH3)2 N(CH2)3 N(CH3)2
MDL Number MFCD00008337
Quantity 500 mL
Synonym 1,3-Bis(dimethylamino)propane; TMPDA
Molecular Weight (g/mol) 130.23
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.