Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-1,3-propanediamine, 99+%

Catalog No. AC155510100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155510100 10 g Glass bottle
AC155510500 50 g Glass bottle
AC155512500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC155510100 Supplier Thermo Scientific Chemicals Supplier No. 155510100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 110-95-2 | C7H18N2 | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-95-2
Molecular Formula C7H18N2
Molecular Weight (g/mol) 130.23
MDL Number MFCD00008337
InChI Key DMQSHEKGGUOYJS-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-1,3-propanediamine, 1,3-bis dimethylamino propane, tetramethyltrimethylenediamine, n1,n1,n3,n3-tetramethylpropane-1,3-diamine, n,n,n',n'-tetramethyl-1,3-diaminopropane, 1,3-propanediamine, n,n,n',n'-tetramethyl, bis dimethylamino methyl methane, n,n,n',n'-tetramethyltrimethylenediamine, unii-73b9i6hn5r
PubChem CID 8084
IUPAC Name N,N,N',N'-tetramethylpropane-1,3-diamine
SMILES CN(C)CCCN(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.7800g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4224 to 1.4244
Linear Formula (CH3)2N(CH2)3N(CH3)2
Quantity 10 g
Beilstein 04, 262
Specific Gravity 0.78
Formula Weight 130.23
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyl-1,3-propanediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth
EINECSNumber 203-818-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.