Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-2,2-dimethyl-1,3-propanediamine 97.0+%, TCI America™
SDP

Catalog No. T100825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T100825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T100825ML Supplier TCI America Supplier No. T100825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53369-79-2
Molecular Formula C9H22N2
Molecular Weight (g/mol) 158.289
MDL Number MFCD00053430
InChI Key MOBAUGYLXJCSAW-UHFFFAOYSA-N
Synonym N,N,N′C,N′C,2,2-Hexamethyl-1,3-propanediamine, N,N,N′C,N′C-Tetramethyl-1,3-diamino-2,2-dimethylpropane, N,N,N′C,N′C-Tetramethylneopentanediamine
PubChem CID 20668853
IUPAC Name N,N,N',N',2,2-hexamethylpropane-1,3-diamine
SMILES CC(C)(CN(C)C)CN(C)C

Specifications

Color Yellow
Boiling Point 162°C
Quantity 25 mL
UN Number 2734
Formula Weight 158.29
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetramethyl-2,2-dimethyl-1,3-propanediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.