Learn More
Medchemexpress LLC N,N,N',N'-tetramethyl-p-phenylenediamine | 100-22-1 | MFCD00008309 | 100.0% | 164.25 g·mol⁻¹ | C10H16N2 | 1 ML
SDP
Catalog No. 5000300113
Encompass_Preferred
N,N,N′,N′-Tetramethyl-p-phenylenediamine (TMPD) is a small organic redox indicator used in biochemical research to probe electron transfer processes, including studies of cellular respiration and peroxidase activity. It is provided as both a solid and as a ready-to-use solution for laboratory experiments.
  • Redox indicator for biochemical and enzymatic assays.
  • Useful probe for assessing impairment of cell respiration.
  • Available as solid and as a 10 mM solution in DMSO for flexible use.
  • High purity suitable for research-grade applications.
  • Molecular formula C10H16N2 and molecular weight 164.25 g·mol⁻¹.
Catalog No. 50-003-00113 Supplier Medchemexpress LLC Supplier No. HYW02135810MM/1ML
Add to Cart
Add to Cart
N,N,N′,N′-Tetramethyl-p-phenylenediamine (TMPD) is a small organic redox indicator used in biochemical research to probe electron transfer processes, including studies of cellular respiration and peroxidase activity. It is provided as both a solid and as a ready-to-use solution for laboratory experiments.
  • Redox indicator for biochemical and enzymatic assays.
  • Useful probe for assessing impairment of cell respiration.
  • Available as solid and as a 10 mM solution in DMSO for flexible use.
  • High purity suitable for research-grade applications.
  • Molecular formula C10H16N2 and molecular weight 164.25 g·mol⁻¹.