Learn More
Medchemexpress LLC N,N,N′,N′-tetramethyl-p-phenylenediamine | 100-22-1 | MFCD00008309 | 100.0% | 164.25 g/mol | C10H16N2 | 10 G
SDP
Catalog No. 5000300940
Encompass_Preferred
N,N,N′,N′-tetramethyl-p-phenylenediamine (TMPD) is an organic redox indicator used in biochemical and analytical assays, commonly applied in studies of cellular respiration and electron transfer. Supplied as a solid reagent for laboratory research use.
  • Redox indicator for biochemical and analytical assays.
  • Useful in studies of cell respiration and electron transfer.
  • High purity suitable for research applications.
  • Solid form with off-white to light brown appearance.
  • Molecular formula C10H16N2; molecular weight 164.25 g/mol.
  • Available in small laboratory pack sizes for convenience.
  • For research use only; not for clinical or human use.
Catalog No. 50-003-00940 Supplier Medchemexpress LLC Supplier No. HYW02135810G
N,N,N′,N′-tetramethyl-p-phenylenediamine (TMPD) is an organic redox indicator used in biochemical and analytical assays, commonly applied in studies of cellular respiration and electron transfer. Supplied as a solid reagent for laboratory research use.
  • Redox indicator for biochemical and analytical assays.
  • Useful in studies of cell respiration and electron transfer.
  • High purity suitable for research applications.
  • Solid form with off-white to light brown appearance.
  • Molecular formula C10H16N2; molecular weight 164.25 g/mol.
  • Available in small laboratory pack sizes for convenience.
  • For research use only; not for clinical or human use.