Learn More
Medchemexpress LLC N,N,N′,N′-tetramethyl-p-phenylenediamine | 100-22-1 | MFCD00008309 | 100.0% | 164.25 g/mol | C10H16N2 | 5 G
SDP
Catalog No. 5000301394
Encompass_Preferred
N,N,N′,N′-Tetramethyl-p-phenylenediamine (TMPD) is a redox indicator and electron donor used as a redox mediator in biochemical and enzymatic assays. It is widely used in oxidase testing and studies of cell respiration, supplied as a research-grade reagent for laboratory use.
  • Redox mediator for biochemical and enzymatic assays.
  • Useful in oxidase tests and cell respiration studies.
  • Acts as an electron donor in spectrophotometric applications.
  • Supplied as a small-scale reagent suitable for research use.
  • High purity appropriate for analytical workflows.
Catalog No. 50-003-01394 Supplier Medchemexpress LLC Supplier No. HYW0213585G
N,N,N′,N′-Tetramethyl-p-phenylenediamine (TMPD) is a redox indicator and electron donor used as a redox mediator in biochemical and enzymatic assays. It is widely used in oxidase testing and studies of cell respiration, supplied as a research-grade reagent for laboratory use.
  • Redox mediator for biochemical and enzymatic assays.
  • Useful in oxidase tests and cell respiration studies.
  • Acts as an electron donor in spectrophotometric applications.
  • Supplied as a small-scale reagent suitable for research use.
  • High purity appropriate for analytical workflows.