Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetramethyl-P-Phenylenediamine Dihydrochloride, Sigma-Aldrich

Catalog No. 01258977 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-977 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-977 Supplier MilliporeSigma Supelco Supplier No. T313425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Redox mediator commonly used for the colorimetric identification of bacteria in the oxidase test.

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride (TMPD) is a redox mediator used in the oxidase test for detecting bacterial cytochrome c oxidases. TMPD is commonly used to perform the oxidase test for the colorimetric identification of pathogenic and non-pathogenic bacteria.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-01-4
Molecular Weight (g/mol) 237.17 g/mol

Specifications

Quantity 25 g
Percent Purity ≥95%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.