Learn More
Medchemexpress LLC N,n,n',n'-Tetramethylchloroformamidinium hexafluorophosphate (TCFH) | 94790-35-9 | 99.63% | 280.58 | 1 KG
SDP
Catalog No. 5000454953
Encompass_Preferred
N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphate (TCFH) is a biochemical coupling reagent. It has applications in medical research, specifically in the study of antitumor drugs.
  • Used as a biological coupling reagent.
  • Applicable in antitumor drug research.
Catalog No. 50-004-54953 Supplier Medchemexpress LLC Supplier No. HYW0115561KG
N,N,N',N'-Tetramethylchloroformamidinium hexafluorophosphate (TCFH) is a biochemical coupling reagent. It has applications in medical research, specifically in the study of antitumor drugs.
  • Used as a biological coupling reagent.
  • Applicable in antitumor drug research.