Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N,N,N'-Trimethylethylenediamine

Catalog No. 12712425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-712-425G 25 g
12-712-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-712-425G Supplier Sigma Aldrich Supplier No. 12712425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n,nα-trimethylethylenediamine is synthesized using laser-vaporization supersonic molecular beam
  • It has been studied using pulsed-field ionization zero electron kinetic energy.n,n,nα-trimethylethylenediamine has been used in the preparation of a novel gemini surfactant, n,n-dimethyl-n-{2-[nα-methyl-nα-(3-sulfopropyl)-alkylammonium]ethyl}-1-alkylammonium bromides (2c(n)amsb, where n represents hydrocarbon chain lengths of 8, 10, 12, and 14), by reacting it with n-alkyl bromide
  • It has also been used as an amine component for in situ formation of α-amino alkoxides

Specifications

CAS 142-25-6
Density 0.786 g/mL (at 25°C (literature))
Boiling Point 116°C to 118°C (lit.)
Molecular Formula C5H14N2
Refractive Index n20/D 1.419 (literature)
Linear Formula (CH3)2 NCH2CH2NHCH3
MDL Number MFCD00014874
Quantity 25 g
Molecular Weight (g/mol) 102.18
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.