Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N'-Trimethylethylenediamine (Tech.), 93%, Thermo Scientific™

Catalog No. AC155620250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC155620250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC155620250 Supplier Thermo Scientific Chemicals Supplier No. 155620250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-25-6
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
InChI Key HVOYZOQVDYHUPF-UHFFFAOYSA-N
Synonym n,n,n'-trimethylethylenediamine, n1,n1,n2-trimethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-trimethyl, 2-dimethylamino ethyl methyl amine, n,n,n-trimethylethylenediamine, ethylenediamine, n,n,n'-trimethyl, n1,n1,n2-trimethyl-1,2-ethanediamine, 1,2-ethanediamine, n1,n1,n2-trimethyl, dimethyl 2-methylamino ethyl amine, n,n,n'-trimethylethane-1,2-diamine
PubChem CID 67338
IUPAC Name N,N',N'-trimethylethane-1,2-diamine
SMILES CNCCN(C)C

Specifications

CAS Min % 91.0
CAS Max % 100.0
Color Colorless
Density 0.8g/mL
Infrared Spectrum Authentic
Assay Percent Range 93%
Refractive Index 1.4185 to 1.4205 (20°C, 589nm)
Linear Formula (CH3)2NCH2CH2NHCH3
Quantity 25 g
Beilstein 04, I, 415
Formula Weight 102.18
Percent Purity ≥91%
Physical Form Liquid
Chemical Name or Material N, N, N'-Trimethylethylenediamine, tech., 93%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.