Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N,N-Trimethylhexadecan-1-aminium-d42 (chloride) | 2095863-88-8 | 99.6% | 362.26 | 1 MG
SDP

Catalog No. 5000366547
Encompass_Preferred

N,N,N-Trimethylhexadecan-1-aminium-d42 (chloride) is a solid chemical product intended for use as a laboratory chemical and in the manufacture of various substances. It is identified by CAS number 2095863-88-8 and has a molecular weight of 362.26.

  • Store at 4°C, sealed, away from moisture.
  • For storage in solvent: -80°C for 6 months, or -20°C for 1 month (sealed storage, away from moisture).
  • Shipping at room temperature is suitable if less than 2 weeks.
  • Not identified as a hazardous substance or mixture.
  • Purity (HPLC) is 99.60%.
  • Isotopic enrichment is 98.97%.
  • Product has been tested and complies with specifications.
  • For research use only; not fully validated for medical applications.

Catalog No. 50-003-66547 Supplier Medchemexpress LLC Supplier No. HYW011291S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N,N-Trimethylhexadecan-1-aminium-d42 (chloride) is a solid chemical product intended for use as a laboratory chemical and in the manufacture of various substances. It is identified by CAS number 2095863-88-8 and has a molecular weight of 362.26.

  • Store at 4°C, sealed, away from moisture.
  • For storage in solvent: -80°C for 6 months, or -20°C for 1 month (sealed storage, away from moisture).
  • Shipping at room temperature is suitable if less than 2 weeks.
  • Not identified as a hazardous substance or mixture.
  • Purity (HPLC) is 99.60%.
  • Isotopic enrichment is 98.97%.
  • Product has been tested and complies with specifications.
  • For research use only; not fully validated for medical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.