Learn More
Medchemexpress LLC Nε,Nε,Nε-trimethyllysine-d9 chloride | 55528-53-5 | 1 MG
SDP
Catalog No. 5000419412
Encompass_Preferred
Nε,Nε,Nε-Trimethyllysine-d9 (chloride) is the deuterium labeled Nε,Nε,Nε-Trimethyllysine (chloride). It serves as a precursor for gut flora-dependent formation of N,N,N-trimethyl-5-aminovaleric acid (TMAVA). This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-19412 Supplier Medchemexpress LLC Supplier No. HYN7404S1MG
Nε,Nε,Nε-Trimethyllysine-d9 (chloride) is the deuterium labeled Nε,Nε,Nε-Trimethyllysine (chloride). It serves as a precursor for gut flora-dependent formation of N,N,N-trimethyl-5-aminovaleric acid (TMAVA). This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS