Learn More
Medchemexpress LLC N,N,N-trimethyloctan-1-aminium chloride | 10108-86-8 | MFCD00059977 | 99.0% | 207.79 g/mol | C11H26ClN | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW014536
Encompass_Preferred
N,N,N-Trimethyloctan-1-aminium chloride is a quaternary ammonium salt commonly used as a cationic surfactant, phase-transfer reagent, and biochemical assay reagent. Supplied as a white to off-white solid with documented purity and supporting SDS/COA, it is intended for laboratory use where stable, high-purity quaternary ammonium salts are required.
  • High purity (99.0%) suitable for analytical and preparative work.
  • White to off-white solid, easy to weigh and store.
  • Functions as a cationic surfactant and phase-transfer catalyst.
  • Compatible with common organic solvents and aqueous systems.
  • Available in multiple package sizes for lab-scale use.
Catalog No. 50-003-03584