Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N'-Triphenyl-1,4-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. T3367200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3367200MG 200 mg
T33671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3367200MG Supplier TCI America Supplier No. T3367200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19606-98-5
Molecular Formula C24H20N2
Molecular Weight (g/mol) 336.44
MDL Number MFCD27939629
InChI Key OCQFHFNWMCLWKC-UHFFFAOYSA-N
Synonym 4-Anilinotriphenylamine
PubChem CID 161288
IUPAC Name N1,N1,N4-triphenylbenzene-1,4-diamine
SMILES N(C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 200 mg
Formula Weight 336.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′-Triphenyl-1,4-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.