Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N'-Triphenylbenzidine 97.0+%, TCI America™
SDP

Catalog No. T28915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28915G 5 g
T28911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28915G Supplier TCI America Supplier No. T28915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 167218-30-6
Molecular Formula C30H24N2
Molecular Weight (g/mol) 412.536
MDL Number MFCD17926476
InChI Key XHPBZHOZZVRDHL-UHFFFAOYSA-N
PubChem CID 22013295
IUPAC Name N-phenyl-4-[4-(N-phenylanilino)phenyl]aniline
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 167°C
Color Yellow
Quantity 5 g
Synonym 4-Diphenylamino-4′-phenylaminobiphenyl
Formula Weight 412.54
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N,N′-Triphenylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.