Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N',N″-Triphenylguanidine, 97%, Thermo Scientific™

Catalog No. AAA1879422
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1879422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1879422 Supplier Thermo Scientific Chemicals Supplier No. A1879422

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-01-9
Molecular Formula C19H17N3
Molecular Weight (g/mol) 287.366
MDL Number MFCD00020648
InChI Key FUPAJKKAHDLPAZ-UHFFFAOYSA-N
Synonym n,n',n-triphenylguanidine, guanidine, n,n',n-triphenyl, sym-triphenylguanidine, guanidine, triphenyl, 1,2,3-trifenylguanidin, unii-i64b170qfg, 1,2,3-trifenylguanidin czech, guanidine, 1,2,3-triphenyl, phenyl 2-phenyl-1-phenylamino-2-azavinyl amine, triphenylguanidin
PubChem CID 7539
IUPAC Name 1,2,3-triphenylguanidine
SMILES C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)NC3=CC=CC=C3

Specifications

Melting Point 142°C to 145°C
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 287.36
Percent Purity 97%
Chemical Name or Material N,N',N''-Triphenylguanidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-907-9
RTECSNumber MF6825000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.