Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N',N″-Tris(p-toluenesulfonyl)diethylenetriamine 98.0+%, TCI America™
SDP

Catalog No. T172325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T172325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T172325G Supplier TCI America Supplier No. T172325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 56187-04-3
Molecular Formula C25H31N3O6S3
Molecular Weight (g/mol) 565.718
MDL Number MFCD00015623
InChI Key OCDIAWYQMCPHAM-UHFFFAOYSA-N
Synonym N,N′C,N′C′C-Tritosyldiethylenetriamine
PubChem CID 316685
IUPAC Name 4-methyl-N-[2-[(4-methylphenyl)sulfonyl-[2-[(4-methylphenyl)sulfonylamino]ethyl]amino]ethyl]benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCCN(CCNS(=O)(=O)C2=CC=C(C=C2)C)S(=O)(=O)C3=CC=C(C=C3)C

Specifications

Melting Point 177°C
Color White
Quantity 25 g
Formula Weight 565.72
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′,N′′-Tris(p-toluenesulfonyl)diethylenetriamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.