Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,Nft.,Nft.-Tetramethylethylenediamine, Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19401950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19401950 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19401950 Supplier MP Biomedicals, Inc Supplier No. 0219401950
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N,N',N'-Tetramethylethylenediamin is a tertiary amine base

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Density 0.775g/mL at 20°C
Assay Percent Range Approx. 99%
Quantity 50 mL
Formula Weight 116.2
Percent Purity 99%
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.