Learn More
Cayman Chemical N,N',N''-Triacetylchitotriose, Purity: ≥95%, CAS: 38864-21-0, 10 mg
SDP
Catalog No. 502980728
Encompass_Preferred
A triose oligosaccharide derived from chitin; scavenges reactive oxygen species and protects DNA from oxidative damage
Catalog No. 50-298-0728 Supplier Cayman Chemical Supplier No. 2075910 MG
A triose oligosaccharide derived from chitin; scavenges reactive oxygen species and protects DNA from oxidative damage