Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N,N'-trans-1,4-cyclohexanediylbis[2-(4-chlorophenoxy)acetamide] | 1597403-47-8 | MFCD27952932 | 99.5% | 451.34 g/mol | C22H24Cl2N2O4 | 10MM 1ML
SDP

Catalog No. 5000235622
Encompass_Preferred

ISRIB (trans-isomer) is a small-molecule inhibitor of the integrated stress response that potently inhibits PERK and reverses eIF2α phosphorylation. It is supplied as a 10 mM solution in DMSO or as solid quantities for use in biochemical and cellular assays; reported purity is 99.49% and molecular weight 451.34 g/mol.

  • Potent PERK inhibition with reported IC50 of 5 nM.
  • Reverses eIF2α phosphorylation in cellular assays.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High purity suitable for research applications.
  • Molecular weight 451.34 g/mol and formula C22H24Cl2N2O4.
  • Useful for biochemical and cell-based studies of the integrated stress response.

Catalog No. 50-002-35622 Supplier Medchemexpress LLC Supplier No. HY1249510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ISRIB (trans-isomer) is a small-molecule inhibitor of the integrated stress response that potently inhibits PERK and reverses eIF2α phosphorylation. It is supplied as a 10 mM solution in DMSO or as solid quantities for use in biochemical and cellular assays; reported purity is 99.49% and molecular weight 451.34 g/mol.

  • Potent PERK inhibition with reported IC50 of 5 nM.
  • Reverses eIF2α phosphorylation in cellular assays.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High purity suitable for research applications.
  • Molecular weight 451.34 g/mol and formula C22H24Cl2N2O4.
  • Useful for biochemical and cell-based studies of the integrated stress response.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.