Learn More
Hach Company N-(1-Naphthyl)-ethylenediamine Dihydrochloride, 25 g
Catalog No. 501994844 Shop All Hach Company Products
Encompass
Reagent for determination of nitrite. Analytical Grade, CAS No. 1465-25-4 C12H14N2·...2HCl, FW = 259.17. Assay: ≥98.0% 25 g bottle
Catalog No. 50-199-4844 Supplier Hach Company Supplier No. 1124224
Reagent for determination of nitrite. Analytical Grade, CAS No. 1465-25-4 C12H14N2·...2HCl, FW = 259.17. Assay: ≥98.0% 25 g bottle