Learn More
Cayman Chemical N NbIs 2 3DIhydroxybenzoyl
SDP
Catalog No. 502990730
Encompass_Preferred
A microbial metabolite and degradation product of enterobactin; inhibits invasion of 26-L5 murine colon cancer cells in a Matrigel™ assay (IC50 = 2.7 µM)
Catalog No. 50-299-0730 Supplier Cayman Chemical Supplier No. 31778500 UG
A microbial metabolite and degradation product of enterobactin; inhibits invasion of 26-L5 murine colon cancer cells in a Matrigel™ assay (IC50 = 2.7 µM)