Learn More
Sigma Organic Chemistry N,N-Diisopropylethylamine | 100ML | 7087-68-5 | MFCD00008868
Supplier: Sigma Organic Chemistry D125806100ML
N,N-Diisopropylethylamine, 100ML
About This Item:
Synonym(s): N-Ethyldiisopropylamine; DIPEA; Ethyldiisopropylamine; 'H++nig's base’
Linear Formula: [(CH3)2CH]2NC2H5
Molecular Weight: 129.24
CAS Number: 7087-68-5
MDL Number: MFCD00008868
UNSPSC Code: 12352100
Purity: >=99%
Boiling Point: 127 C (lit.)
Melting Point: <-50 C (lit.)
Density: 0.742 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.414 (lit.)
Storage: room temp
EINECS Number: 230-392-0
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.