Learn More
Cayman Chemical N,N-Dimethylcathinone (hydrochloride), Purity: ≥98%, CAS: 10105-90-5, 50 mg
SDP
Catalog No. 502894318
Encompass_Preferred
A psychotropic compound of the phenethylamine and cathinone classes; slightly less potent than methcathinone and essentially equipotent with cathinone, in behavioral studies involving rats; intended for forensic purposes
Catalog No. 50-289-4318 Supplier Cayman Chemical Supplier No. 900114450 MG
A psychotropic compound of the phenethylamine and cathinone classes; slightly less potent than methcathinone and essentially equipotent with cathinone, in behavioral studies involving rats; intended for forensic purposes