Learn More
Cayman Chemical N,N'-Diphenyl-1,4-phenylenediamine, Purity: ≥98%, CAS: 74-31-7, 100 g
SDP
Catalog No. 502944729
Encompass_Preferred
A p-phenylenediamine and non-oxidized version of DPPD-Q; toxic to the aquatic bacterium V. fischeri (EC50 = 0.02 mg/L); reduces the viability of HepG2 cancer cells at 10 µg/L; decreases urinary protein levels and serum levels of bilirubin in male rats at 100 mg/kg per day; increases the time in gestation for female rats when administered to pregnant dams at 50 and 300 mg/kg per day; has been found in water treatment plant influent, biosolids, and landfill leachates, indoor dust, and in PM2.5 samples collected in cities and by roadsides
Catalog No. 50-294-4729 Supplier Cayman Chemical Supplier No. 40727100 G
A p-phenylenediamine and non-oxidized version of DPPD-Q; toxic to the aquatic bacterium V. fischeri (EC50 = 0.02 mg/L); reduces the viability of HepG2 cancer cells at 10 µg/L; decreases urinary protein levels and serum levels of bilirubin in male rats at 100 mg/kg per day; increases the time in gestation for female rats when administered to pregnant dams at 50 and 300 mg/kg per day; has been found in water treatment plant influent, biosolids, and landfill leachates, indoor dust, and in PM2.5 samples collected in cities and by roadsides