Learn More
Medchemexpress LLC N-nervonoyl taurine | 1942003-30-6 | 99.0% | 473.75 | C26H51NO4S | 1mg
SDP
Catalog No. 5000269288
Encompass_Preferred
N-Nervonoyl taurine ((Z)-2-tetracos-15-enamidoethanesulfonic acid) is a fatty acid-taurine conjugate derived from nervonic acid N-Nervonoyl taurine is a substrate of fatty acid amide hydrolase (FAAH) discovered during metabolite profiling[1]
Catalog No. 50-002-69288 Supplier Medchemexpress LLC Supplier No. HYW7514181MG
N-Nervonoyl taurine ((Z)-2-tetracos-15-enamidoethanesulfonic acid) is a fatty acid-taurine conjugate derived from nervonic acid N-Nervonoyl taurine is a substrate of fatty acid amide hydrolase (FAAH) discovered during metabolite profiling[1]