Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,Nft.-Octamethylenebis(2,2-dichloroacetamide), 98.2%, MP Biomedicals™

Catalog No. ICN15805083 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15805083 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15805083 Supplier MP Biomedicals, Inc Supplier No. 0215805083
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1477-57-2
Molecular Formula C12H20Cl4N2O2
Molecular Weight (g/mol) 366.104
MDL Number MFCD00000841
InChI Key FAOMZVDZARKPFJ-UHFFFAOYSA-N
Synonym fertilysin, bis-diamine, n,n'-octane-1,8-diyl bis 2,2-dichloroacetamide, fertilysine, n,n'-octamethylenebis dichloroacetamide, bisdiamine, fertilysine n, n,n'-bis dichloroacetyl-1,8-octamethylenediamine, n,n'-octamethylenebis 2,2-dichloroacetamide, n,n'-bis dichloracetyl-1,8-octanediamine
PubChem CID 15134
IUPAC Name 2,2-dichloro-N-[8-[(2,2-dichloroacetyl)amino]octyl]acetamide
SMILES C(CCCCNC(=O)C(Cl)Cl)CCCNC(=O)C(Cl)Cl

Specifications

Color White
Assay Percent Range 98.2%
Quantity 250 mg
Formula Weight 366.11
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N, N'-Octamethylenebis(2, 2-dichloroacetamide)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.